Learn More
Ace Glass, Inc. Immersion well, outer chamber only, 220mm
SDP
Catalog No. 50979574
Encompass
220mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.
Catalog No. 50-979-574 Supplier Ace Glass, Inc. Supplier No. 785807
220mm quartz immersion well, triple-walled, designed for low temperature reactions cooled up to -78 degrees C. The outer two walls are permanently sealed and evacuated. The inner removable tube set consists of one each quartz and borosilicate tubes.