Learn More
Electron Microscopy Sciences Immersion Cedar Wood Oil 100 ML
SDP
Catalog No. 50980291
Encompass_Preferred
Refractive Index 20 degree C 1.515-1.520. Used as an immersion oil.
Catalog No. 50-980-291 Supplier Electron Microscopy Sciences Supplier No. 12430
Refractive Index 20 degree C 1.515-1.520. Used as an immersion oil.