Learn More
BURKLE INC Immersion cylinder, brass, 33.81 oz. (1000 ml), hxØ 450x75 mm
Catalog No. 5000025872
Encompass_Preferred
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants in Explosion Categories IIA, IIB and IIC. Completely made from non-spark producing materials, nickel-plated.
Catalog No. 50-000-25872 Supplier BURKLE INC Supplier No. 53657000
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants in Explosion Categories IIA, IIB and IIC. Completely made from non-spark producing materials, nickel-plated.