Learn More
BURKLE INC Immersion cylinder, brass, 16.91 oz. (500 ml), hxØ 300x75 mm
Catalog No. 5000025565
Encompass_Preferred
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants in Explosion Categories IIA, IIB and IIC. Completely made from non-spark producing materials, nickel-plated.
Catalog No. 50-000-25565 Supplier BURKLE INC Supplier No. 53657500
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum and liquid lubricants in Explosion Categories IIA, IIB and IIC. Completely made from non-spark producing materials, nickel-plated.