Learn More
Camag Scientific Chromatogram Immersion Device 3, For Tlc And Hptlc Plates Up To 20 X 20 Cm, Without Dip Tanks
SDP
Catalog No. 501979320
Encompass_Preferred
The Chromatogram Immersion Device 3 Is Used For Automated Dipping. Tlc/Hptlc Plates Are Immersed With Uniform Vertical Speed Into Derivatization Reagents Or Solutions For Plate Impregnation.
Catalog No. 50-197-9320 Supplier Camag Scientific Supplier No. 022.6606
The Chromatogram Immersion Device 3 Is Used For Automated Dipping. Tlc/Hptlc Plates Are Immersed With Uniform Vertical Speed Into Derivatization Reagents Or Solutions For Plate Impregnation.