Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Immersion Heated, Mobile Self-Contained Emergency Safety Shower

Catalog No. 18666709 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Green
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-666-709 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-666-709 Supplier Justrite Manufacturing Co Supplier No. MHW20001
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Hughes Immersion Heated, Mobile Self-Contained Emergency Safety Shower features a 528-gallon (2000-L) tank for reliable, effective treatment to accident victims in remote locations where a water supply is unavailable

Features:

  • Durable, chemical-resistant polyethylene tank incorporates an electric submersible pump.
  • Constructed from a durable, corrosion-resistant acrylic capped ABS, the eye/face wash incorporates an integral lid connected to the water valve for automatic operation—activate by simply pulling down the lid.
  • Easy-to-reach flow regulator valve positioned on side of bowl adjusts to suit the severity of the accident.
  • Includes heavy-duty galvanized trailer with toe hitch featuring four braked wheels with standard trailer-type pneumatic tires for easy movement between locations.
  • Includes a universal safety shower and eyewash sign for enhanced visibility.
  • Suitable for use in non-hazardous areas.
  • Electrical supply: 110V Single Phase, 60/50Hz
  • Mobile eye/face wash and shower provides a continuous flow of tepid water for over 15 minutes
  • Covered bowl ensures that the nozzles and bowl remain clean and free from contamination
  • High-performance quarter-turn ball valves made of 316L stainless steel endure the harshest environments and site conditions
  • Integral immersion heater, controlled by a dual safety thermostat, maintains the water in the tank at a constant temperature
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty and Services

1 Year

Specifications

Capacity (English) 528 gal.
Color Green
Depth (English) 5 in.
Depth (Metric) 127 mm
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 5 in.
Height (Metric) 127 mm
Includes 1 Unit
Material Ceramic Media
Product Type Safety Shower and Emergency Eye Wash
Width (English) 33 in.
Width (Metric) 838 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.