Learn More
Cole-Parmer Meiji Techno MA700 Microscopy Immersion Oil, 15 mL
Supplier:  Cole-Parmer 4840195
Encompass
Type A immersion oil has a viscosity of 150 cSt and a refractive index of 1.5239. Use with compound microscopes that have a 100x immersion objective.
Catalog No. NC0616187