Learn More
Azzota Corp Immersion Oil For Any Microscopes, Refractive Index Nd=1.515 , 7 Ml, Standardized At 23'C
Catalog No. 507532972
Encompass_Preferred
Immersion Oil for any microscopes, Refractive index ND=1.515 , 7 Ml, Standardized at 23'C
Catalog No. 50-753-2972 Supplier Azzota Corp Supplier No. MOIL
Immersion Oil for any microscopes, Refractive index ND=1.515 , 7 Ml, Standardized at 23'C