Learn More
G-Biosciences Immersion Oil
SCI_ED
Catalog No. S000757
Change view
Click to view available options
Quantity:
15 mL
Catalog No. Quantity
S000757 15 mL
1 options
Catalog No. S000757 Supplier G-Biosciences Supplier No. 7862206
Add to Cart
Add to Cart
Immersion oil is a clear, viscous liquid with optimized refractive properties, specifically modified to closely approximate the refractive index (RI) of glass (ne = 1.5). It is used in conjunction with an objective lens to enhance the resolving power.
It is applied dropwise to stained and mounted or non-mounted specimen material to form a clear film between the specimen and the microscope lens to eliminate the deflection of incident light and thus substantially enhance the optical efficiency of the lens.
Specifications
Type Immersion Oil
Viscosity High
Quantity 15 mL