Learn More
THORLABS INC Low Autofluorescence Immersion Oil, n = 1.518, Olympus Type F, 30 mL
Catalog No. NC0297589
Encompass
Low Autofluorescence Immersion Oil, n = 1.518, Olympus Type F, 30 mL
Catalog No. NC0297589 Supplier THORLABS INC Supplier No. MOIL30
Low Autofluorescence Immersion Oil, n = 1.518, Olympus Type F, 30 mL