Learn More
Electron Microscopy Sciences Cargille Microscope Immersion Oil Type 300 16 OZ
SDP
Supplier:  Electron Microscopy Sciences 1691716
Encompass_Preferred
Cargille PCB-Free Immersion oils were introduced in 1972. Today, no other immersion oil meets FDA criteria.  Type 300 meets the stringent requirements of Automated Hematology Systems including specialized viscosity and exacting controls for its consistency.
Density: @ 23°C: .923 g/cc
Catalog No. 50-366-046