Learn More
Electron Microscopy Sciences Cargille Microscope Immersion Oil Type A 120 ML
SDP
Supplier:  Electron Microscopy Sciences 16909
Encompass_Preferred
Cargille PCB-Free Immersion oils were introduced in 1972. Today, no other immersion oil meets FDA criteria.  Viscosity 150 cSt for normal light microscopy.
Density: @ 23°C: 0.918 g/cc
Catalog No. 50-266-37