Learn More
Electron Microscopy Sciences Cargille Immersion Oil Type FF 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 1691601
Encompass_Preferred
After years of development and field testing, we now have available two new PCB-Free Immersion Oils for Fluorescence Microscopy. Has virtually no background fluorescence making it ideal for observing low levels of fluorescence. Highly stable, water white, and non-hydroscopic. Substituted for other fluorescence microscopy immersion oils.
Catalog No. 50-331-26