Learn More
Electron Microscopy Sciences Cargille Immersion Oil Type HF 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 1691501
Encompass_Preferred
After years of development and field testing, we now have available two new PCB-Free Immersion Oils for Fluorescence Microscopy. A new Immersion Oil called HF which would provide low fluorescence and the correct optical properties.
Catalog No. 50-331-23