Learn More
Electron Microscopy Sciences Cargille Microscope Immersion Oil Type NVH Oil 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 1690901
Encompass_Preferred
Cargille PCB-Free Immersion oils were introduced in 1972. Today, no other immersion oil meets FDA criteria.  For horizontal, inverted and inclined instruments and projection equipment where higher viscosities are generally required.   Type NVH meets the requirement, having a viscosity of 21,000cST.
Density: @ 23°C: 0.919 g/cc
Catalog No. 50-266-40