Learn More
Thermco Products Inc Tissue Flotation Bath Thermometer, -15/105Deg.C x 1.0Deg.C, TOTAL Imm, Spirit
Catalog No. NC0731987
Encompass_Preferred
Non-mercury thermometer used in tissue flotation bath to monitor the temperature. The ACC535S1 thermometer lies on the bottom of the bath. It has a rectangular ring attached, so it will not roll. The ring also allows the thermometer scale to face up, so the scale is easy to read the correct temperature. The red liquid in the thermometer is easy to see through the bath liquid
Catalog No. NC0731987 Supplier Thermco Products Inc Supplier No. ACC535S1
Non-mercury thermometer used in tissue flotation bath to monitor the temperature. The ACC535S1 thermometer lies on the bottom of the bath. It has a rectangular ring attached, so it will not roll. The ring also allows the thermometer scale to face up, so the scale is easy to read the correct temperature. The red liquid in the thermometer is easy to see through the bath liquid