Learn More
Medchemexpress LLC Immh-010 maleate | 2541982-47-0 | 99.8% | 756.04 | 5 MG
SDP
Catalog No. 5000393625
Encompass_Preferred
IMMH-010 maleate is a prodrug that acts as a novel PD-L1 inhibitor, showing potential antitumor activity. It is being investigated for the treatment of neurological disorders and advanced malignant solid tumors. Upon oral administration, it quickly converts into its active form, YPD-29B. This compound is significant for advancing research in the field of PD-L1 inhibitors and related therapeutic applications.
  • Acts as a novel PD-L1 inhibitor
  • Shows potential antitumor activity
  • Investigated for neurological disorders and advanced malignant solid tumors
  • Converts into active form, YPD-29B, upon oral administration
  • Advances research in PD-L1 inhibitors and related therapeutic applications
Catalog No. 50-003-93625 Supplier Medchemexpress LLC Supplier No. HY1678405MG
IMMH-010 maleate is a prodrug that acts as a novel PD-L1 inhibitor, showing potential antitumor activity. It is being investigated for the treatment of neurological disorders and advanced malignant solid tumors. Upon oral administration, it quickly converts into its active form, YPD-29B. This compound is significant for advancing research in the field of PD-L1 inhibitors and related therapeutic applications.
  • Acts as a novel PD-L1 inhibitor
  • Shows potential antitumor activity
  • Investigated for neurological disorders and advanced malignant solid tumors
  • Converts into active form, YPD-29B, upon oral administration
  • Advances research in PD-L1 inhibitors and related therapeutic applications