Learn More
Proteochem Inc Immobilized Chymotrypsin 2 ml
Catalog No. NC1615811
Encompass_Preferred
Immobilized Chymotrypsin Resin (Chymotrypsin Agarose) is used to preferentially hydrolyze proteins at Tyrosine, Tryptophan, and Phenylalanine residues with minimal Chymotrypsin autolysis byproducts or other protease contaminants. Immobilized Chymotrypsin resin is provided in a 50% slurry, and is supplied as 2 mL of actual resin (4 mL of slurry).
Catalog No. NC1615811 Supplier Proteochem Inc Supplier No. G41052ML
Immobilized Chymotrypsin Resin (Chymotrypsin Agarose) is used to preferentially hydrolyze proteins at Tyrosine, Tryptophan, and Phenylalanine residues with minimal Chymotrypsin autolysis byproducts or other protease contaminants. Immobilized Chymotrypsin resin is provided in a 50% slurry, and is supplied as 2 mL of actual resin (4 mL of slurry).