Learn More
Abcam Immobilized Protein G Agarose Beads, 50UL
Catalog No. 502673265
Encompass_Preferred
Protein G Agarose is useful for affinity purification of IgG from many mammalian species.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-3265 Supplier Abcam Supplier No. AB17481650UL
Protein G Agarose is useful for affinity purification of IgG from many mammalian species.
The product is subject to the following: Abcam Restricted Use Statement