Learn More
New England Biolabs, Inc. Immobilized T4 DNA Ligase - 1.1 mg
SDP
Catalog No. 502108550
Encompass
Immobilized T4 DNA Ligase (IM-T4 DNA Ligase) is a slurry of magnetic beads coated with T4 DNA Ligase to produce a 10 mg/ml solution (50% glycerol) with an effective concentration of 60 cohesive end units (CEU) per microliter of slurry. Following a reaction, enzyme can be removed using a magnet thereby avoiding heat inactivation, and can be re-used.
Catalog No. 50-210-8550 Supplier New England Biolabs, Inc. Supplier No. M0569S
Immobilized T4 DNA Ligase (IM-T4 DNA Ligase) is a slurry of magnetic beads coated with T4 DNA Ligase to produce a 10 mg/ml solution (50% glycerol) with an effective concentration of 60 cohesive end units (CEU) per microliter of slurry. Following a reaction, enzyme can be removed using a magnet thereby avoiding heat inactivation, and can be re-used.