Learn More
Meridian Life Science IMMOMIX RED 500RXNS
SDP
Catalog No. NC9874911
Encompass
ImmoMix Red is a complete ready-to-go heat activated 2x reaction mix for carrying out primer extensions which only requires the user to add water, template and primers, and then to pre-heat to 95 C.
Catalog No. NC9874911 Supplier Meridian Life Science Supplier No. BIO25022
ImmoMix Red is a complete ready-to-go heat activated 2x reaction mix for carrying out primer extensions which only requires the user to add water, template and primers, and then to pre-heat to 95 C.