Learn More
Vector Laboratories ImmPACT NovaRED™ Peroxidase (HRP) Substrate Kit 120 ml
SDP
Catalog No. NC9925963
Encompass
ImmPACT NovaRED™ Peroxidase (HRP) Substrate produces an intense red reaction product, with 2-4X the sensitivity of standard VECTOR NovaRED substrate. Sections stained with NovaRED can be dehydrated, cleared, and permanently mounted. This chromogen is useful as an alternative to DAB or as a second color for multiple antigen labeling. ImmPACT NovaRED Substrate also provides excellent color contrast in pigmented tissue such as melanoma and can be used for both manual and automated staining methods. ImmPACT NovaRED™ substrate produces a red reaction product for use in immunohistochemical applications. This product generates 120 ml of working solution that is stable for up to 14 days when stored at 4 degrees C.
Catalog No. NC9925963 Supplier Vector Laboratories Supplier No. SK4805
ImmPACT NovaRED™ Peroxidase (HRP) Substrate produces an intense red reaction product, with 2-4X the sensitivity of standard VECTOR NovaRED substrate. Sections stained with NovaRED can be dehydrated, cleared, and permanently mounted. This chromogen is useful as an alternative to DAB or as a second color for multiple antigen labeling. ImmPACT NovaRED Substrate also provides excellent color contrast in pigmented tissue such as melanoma and can be used for both manual and automated staining methods. ImmPACT NovaRED™ substrate produces a red reaction product for use in immunohistochemical applications. This product generates 120 ml of working solution that is stable for up to 14 days when stored at 4 degrees C.