Learn More
Vector Laboratories ImmPRESS Excel Mouse HRP, 50ml MP-7800-15
SDP
Catalog No. 502045690
Encompass
ImmPRESS Excel Amplified Polymer Staining Kit, Anti-Mouse IgG, Peroxidase, 50 ml, is an enzymatic amplification system that produces crisp, highly sensitive, specific staining with low background. It capitalizes on the advantages of the ImmPRESS polymerized reporter enzyme staining system technology but offers additional sensitivity and convenience. This kit employs a ready-to-use (RTU) Amplifier Antibody followed by RTU ImmPRESS Excel HRP Polymer Reagent, both of which are affinity-purified and extensively cross-adsorbed. The ImmPRESS Excel Reagent is conjugated to HRP micropolymers, a novel approach that avoids the use of large dextrans or other macromolecules as a backbone and allows a higher density of enzymes per antibody to bind the target with minimal interference. The result is an increase in target accessibility, signal intensity, and significantly less background staining. The included ImmPACT DAB EqV Peroxidase (HRP) Substrate produces a crisp, dark brown reaction product.
Catalog No. 50-204-5690 Supplier Vector Laboratories Supplier No. MP780015
ImmPRESS Excel Amplified Polymer Staining Kit, Anti-Mouse IgG, Peroxidase, 50 ml, is an enzymatic amplification system that produces crisp, highly sensitive, specific staining with low background. It capitalizes on the advantages of the ImmPRESS polymerized reporter enzyme staining system technology but offers additional sensitivity and convenience. This kit employs a ready-to-use (RTU) Amplifier Antibody followed by RTU ImmPRESS Excel HRP Polymer Reagent, both of which are affinity-purified and extensively cross-adsorbed. The ImmPRESS Excel Reagent is conjugated to HRP micropolymers, a novel approach that avoids the use of large dextrans or other macromolecules as a backbone and allows a higher density of enzymes per antibody to bind the target with minimal interference. The result is an increase in target accessibility, signal intensity, and significantly less background staining. The included ImmPACT DAB EqV Peroxidase (HRP) Substrate produces a crisp, dark brown reaction product.