Learn More
Vector Laboratories ImmPRESS-HRP Anti-Mouse IgG (Horseradish Peroxidase) Polymer Detection Kit, Made In Goat 50 ml
SDP
Catalog No. NC1119521
Encompass
The ImmPRESS-HRP Anti-Mouse IgG (Peroxidase) Polymer Detection Kit is made in goat and targets mouse primary antibodies. This IHC staining system uses novel conjugation chemistry to create a highly sensitive, ready-to-use, one-step, biotin-free detection system. (A combination of highly active enzyme (peroxidase or alkaline phosphatase) and affinity-purified secondary antibodies results in crisp, strong staining. The ImmPRESS™ micropolymer technology minimizes steric interference and provides enhanced accessibility to the primary antibody targets. The IHC immunostaining protocol is simple. Following a blocking step with 2.5% goat serum, sections are incubated with primary antibody. After a brief wash, the appropriate ImmPRESS™ Reagent is added to the sections and incubated for 30 minutes. Sections are again rinsed and the slides are developed with a peroxidase or alkaline phosphatase substrate. The ImmPRESS Reagent is ready-to-use and stains 250-500 sections.
Catalog No. NC1119521 Supplier Vector Laboratories Supplier No. MP745250
The ImmPRESS-HRP Anti-Mouse IgG (Peroxidase) Polymer Detection Kit is made in goat and targets mouse primary antibodies. This IHC staining system uses novel conjugation chemistry to create a highly sensitive, ready-to-use, one-step, biotin-free detection system. (A combination of highly active enzyme (peroxidase or alkaline phosphatase) and affinity-purified secondary antibodies results in crisp, strong staining. The ImmPRESS™ micropolymer technology minimizes steric interference and provides enhanced accessibility to the primary antibody targets. The IHC immunostaining protocol is simple. Following a blocking step with 2.5% goat serum, sections are incubated with primary antibody. After a brief wash, the appropriate ImmPRESS™ Reagent is added to the sections and incubated for 30 minutes. Sections are again rinsed and the slides are developed with a peroxidase or alkaline phosphatase substrate. The ImmPRESS Reagent is ready-to-use and stains 250-500 sections.