Learn More
Vector Laboratories ImmPRESS-HRP Universal Antibody IgG (Anti-Mouse IgG/Anti-Rabbit IgG, Peroxidase) Polymer Detection Kit, Made In Horse 15 ml
SDP
Supplier:  Vector Laboratories MP750015
Encompass
This ImmPRESS-HRP Universal Antibody IgG (Anti-Mouse IgG/Anti-Rabbit IgG, Peroxidase) Polymer Detection Kit is made in horse and targets both mouse and rabbit primary antibodies. This allows simplified sample processing when both types of primary antibodies are used in a lab. The ImmPRESS reporter enzyme staining system uses novel conjugation chemistry to create a highly sensitive, ready-to-use, one-step, biotin-free detection system. A combination of highly active enzyme (peroxidase or alkaline phosphatase) and affinity-purified secondary antibodies results in crisp, strong staining. The IHC immunostaining protocol is simple: Following a blocking step with 2.5% horse serum, sections are incubated with primary antibody. After a wash, the ImmPRESS Reagent is added to the sections and incubated for 30 minutes. Sections are again rinsed and the slides are developed with a peroxidase or alkaline phosphatase substrate. The ImmPRESS Reagent is ready-to-use and stains 75-150 sections.
Catalog No. NC0271541