Learn More
Vector Laboratories ImmPRESS-HRP Universal PLUS MP-6402-15
SDP
Catalog No. 502045688
Encompass
ImmPRESS-HRP Universal (Horse Anti-Mouse/Rabbit IgG) PLUS Polymer Kit, Peroxidase, 1 Kit, is a polymerized reporter enzyme staining system that uses novel conjugation and micropolymer chemistries to attach highly active enzyme to affinity-purified secondary antibodies. Unlike other polymer systems that use large dextrans or other macromolecules as backbones, the peroxidase micropolymers of the ImmPRESS-HRP Universal (Horse Anti-Mouse/Rabbit IgG) PLUS Polymer Kit limit steric interference for enhanced accessibility to the target. The result is crisp, strong staining, especially of nuclear and membrane antigens, such as Ki67, estrogen receptor, bcl-2, CD3, CD8 and CD10, and greater sensitivity than other polymer systems, in a ready-to-use, one-step detection kit. This kit should not be used to stain tissues from rodents or rabbits due to its reactivity with IgG present in the tissues of these species. The 15 ml kit will stain approximately 75–150 sections.
Catalog No. 50-204-5688 Supplier Vector Laboratories Supplier No. MP640215
ImmPRESS-HRP Universal (Horse Anti-Mouse/Rabbit IgG) PLUS Polymer Kit, Peroxidase, 1 Kit, is a polymerized reporter enzyme staining system that uses novel conjugation and micropolymer chemistries to attach highly active enzyme to affinity-purified secondary antibodies. Unlike other polymer systems that use large dextrans or other macromolecules as backbones, the peroxidase micropolymers of the ImmPRESS-HRP Universal (Horse Anti-Mouse/Rabbit IgG) PLUS Polymer Kit limit steric interference for enhanced accessibility to the target. The result is crisp, strong staining, especially of nuclear and membrane antigens, such as Ki67, estrogen receptor, bcl-2, CD3, CD8 and CD10, and greater sensitivity than other polymer systems, in a ready-to-use, one-step detection kit. This kit should not be used to stain tissues from rodents or rabbits due to its reactivity with IgG present in the tissues of these species. The 15 ml kit will stain approximately 75–150 sections.