Learn More
Vector Laboratories ImmPRESS-AP Anti-Rat,M adsorb. MP-6401-15
SDP
Catalog No. 502045687
Encompass
ImmPRESS-AP Goat Anti-Rat IgG (Mouse Adsorbed) Polymer Detection Kit, Alkaline Phosphatase, 15 ml, is a polymerized reporter enzyme staining system that uses novel conjugation and micropolymer chemistries to attach highly active enzyme to affinity-purified secondary antibodies. Unlike other polymer systems that use large dextrans or other macromolecules as backbones, the alkaline phosphatase micropolymers of the ImmPRESS-AP Goat Anti-Rat IgG (Mouse Adsorbed) Polymer Detection Kit limit steric interference and provide enhanced accessibility to the target. The result is crisp, strong staining, especially of nuclear and membrane antigens, and greater sensitivity than conventional alkaline phosphatase antibody conjugates and other alkaline phosphatase polymer systems, in a ready-to-use, one-step detection kit. The 15 ml kit will stain approximately 75–150 sections.
Catalog No. 50-204-5687 Supplier Vector Laboratories Supplier No. MP640115
ImmPRESS-AP Goat Anti-Rat IgG (Mouse Adsorbed) Polymer Detection Kit, Alkaline Phosphatase, 15 ml, is a polymerized reporter enzyme staining system that uses novel conjugation and micropolymer chemistries to attach highly active enzyme to affinity-purified secondary antibodies. Unlike other polymer systems that use large dextrans or other macromolecules as backbones, the alkaline phosphatase micropolymers of the ImmPRESS-AP Goat Anti-Rat IgG (Mouse Adsorbed) Polymer Detection Kit limit steric interference and provide enhanced accessibility to the target. The result is crisp, strong staining, especially of nuclear and membrane antigens, and greater sensitivity than conventional alkaline phosphatase antibody conjugates and other alkaline phosphatase polymer systems, in a ready-to-use, one-step detection kit. The 15 ml kit will stain approximately 75–150 sections.