Learn More
Andwin Scientific IMMULON 4 PLATE
SDP
Catalog No. NC9939836
Encompass
The Immulon 4 HBXExtra High Binding plate has an uncommon resin that provides maximum protein uptake with greater precision and ensured coefficient of variation (CV) of <5%. This product has a flat-bottom well shape and a well volume capacity of 0.33 mL. There are 96 wells. This product is sold in a pack of 50.
Catalog No. NC9939836 Supplier Andwin Scientific Supplier No. D17506
The Immulon 4 HBXExtra High Binding plate has an uncommon resin that provides maximum protein uptake with greater precision and ensured coefficient of variation (CV) of <5%. This product has a flat-bottom well shape and a well volume capacity of 0.33 mL. There are 96 wells. This product is sold in a pack of 50.