Learn More
Chondrex Inc IMMUN BOVINE COL2, PDR, 10 MG
Catalog No. 501526983
Encompass_Preferred
10 mg (lyophilized) of bovine type II collagen purified from bovine articular cartilage. Purity: >99% by SDS-PAGE analysis (free of pepsin and proteoglycans). Molecular Weight: 300 kDa. Intended for use as antigen for immunizing animals in the collagen-induced arthritis (CIA) model, a standard for collagen analysis by SDS-gel, or substrate for collagenase activity.
Catalog No. 50-152-6983 Supplier Chondrex Inc Supplier No. 20021
10 mg (lyophilized) of bovine type II collagen purified from bovine articular cartilage. Purity: >99% by SDS-PAGE analysis (free of pepsin and proteoglycans). Molecular Weight: 300 kDa. Intended for use as antigen for immunizing animals in the collagen-induced arthritis (CIA) model, a standard for collagen analysis by SDS-gel, or substrate for collagenase activity.