Learn More
Chondrex Inc Immunization Grade Chick Type II Collagen, Solution, 2 mg/mL x 5 mL
Catalog No. 501526981
Encompass_Preferred
10 mg (lyophilized) of chicken type II collagen purified from chicken sternal cartilage. Purity: >99% by SDS-PAGE analysis (free of pepsin and proteoglycans). Molecular Weight: 300 kDa. Intended for use as antigen for immunizing animals in the collagen-induced arthritis (CIA) model, a standard for collagen analysis by SDS-gel, or substrate for collagenase activity.
Catalog No. 50-152-6981 Supplier Chondrex Inc Supplier No. 20011
10 mg (lyophilized) of chicken type II collagen purified from chicken sternal cartilage. Purity: >99% by SDS-PAGE analysis (free of pepsin and proteoglycans). Molecular Weight: 300 kDa. Intended for use as antigen for immunizing animals in the collagen-induced arthritis (CIA) model, a standard for collagen analysis by SDS-gel, or substrate for collagenase activity.