Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Immune Clues : Diagnosing Allergic Reactions
SDPSCI_ED

Catalog No. S001098 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S001098 9+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001098 Supplier EDVOTEK™ Supplier No. 260
Add to Cart
Add to Cart

In immune clues students will review a patient’s symptoms and meal history, perform a simulated skin prick allergy test to identify possible triggers, and use a component-resolved diagnostic simulated blood test to distinguish between a true food allergy and cross-reactivity.

  • Act as an allergist to investigate a patient's allergic reaction.
  • Review the patient's symptoms, test results, and meal history.
  • Perform a simulated skin prick test to check for possible allergens.
  • Use a simulated blood allergen testing to confirm if the reaction is a true food allergy or cross-reactivity with pollen.
  • Group Size: For 10 Lab Groups.
  • Kit Includes: Instructions, samples, antigens and antibodies, various solutions and reagents, pipets and microtest tubes all you need: automatic micropipettes with tips, laboratory glassware, distilled or deionized water, microwave.
  • All You Need: Distilled or Deionized Water, Beakers (250 mL), Pipet Pump or Bulb, Disposable Lab Gloves, Safety Goggles, Automatic Micropipettes (5-50μL, 100-1000μL) and tips.
  • Storage: Some Components Require Refrigerator Storage.

Specifications

Grade Level High School, Higher Ed
Grade 9+
Class Size 10 Lab Groups
Product Type Lab Investigation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.