Learn More
Creative Biomart Scorpion (centruroides) immune Fab2 antivenin (equine)
Catalog No. 502084220
Encompass_Preferred
The product is a purified preparation of immune globulin F(ab)2 fragments that are derived from plasma of the horses immunized with venom of C. noxius, C. l. limpidus, C. l. tecomanus, and C. s. suffusus. It is produced from equine IgG antibodies.
Catalog No. 50-208-4220 Supplier Creative Biomart Supplier No. THP0169
The product is a purified preparation of immune globulin F(ab)2 fragments that are derived from plasma of the horses immunized with venom of C. noxius, C. l. limpidus, C. l. tecomanus, and C. s. suffusus. It is produced from equine IgG antibodies.