Learn More
Creative Biomart Equine Botulinum Neurotoxin F Immune FAB2
Catalog No. 502084218
Encompass_Preferred
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype F.
Catalog No. 50-208-4218 Supplier Creative Biomart Supplier No. THP0167
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype F.