Learn More
Creative Biomart Equine Botulinum Neurotoxin A Immune FAB2
Catalog No. 502084213
Encompass_Preferred
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype A.
Catalog No. 50-208-4213 Supplier Creative Biomart Supplier No. THP0162
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype A.