Learn More
Creative Biomart Equine Botulinum Neurotoxin B Immune FAB2
Catalog No. 502084214
Encompass_Preferred
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype B.
Catalog No. 50-208-4214 Supplier Creative Biomart Supplier No. THP0163
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype B.