Learn More
Creative Biomart Equine Botulinum Neurotoxin E Immune FAB2
Catalog No. 502084217
Encompass_Preferred
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype E.
Catalog No. 50-208-4217 Supplier Creative Biomart Supplier No. THP0166
The product is composed of a mixture of immune globulin fragments purified from plasma of horses that were previously immunized with botulinum toxin serotype E.