Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Breakable Modules Clear
GREENER_CHOICE

Catalog No. 21377265 Shop All Thermo Scientific Products
Change view
Click to view available options
Binding Property:
Hydrophilic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Binding Property
21-377-265 Hydrophilic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-377-265 Supplier Thermo Scientific™ Supplier No. 95029180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.

Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
  • For solid phase immuno assays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fit standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Order Info

dimVolumeMetricWell

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Hydrophilic
Certified Yes
Format Strip Plate
Lid Without Lid
Description High optical quality for reliable results
Detection Method Colorimetric
Packaging 25 Pk., 50 Cases
Color Clear
Product Type Strip Plate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) For antibody sandwich assays
No. per Case 50
No. per Pack 50
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F8
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.