Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Catalog No. 14245211 Shop All Thermo Scientific Products
Change view
Click to view available options
Surface Treatment:
Immulon 2 HB
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
14-245-211 Immulon 2 HB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-245-211 Supplier Thermo Scientific™ Supplier No. 6506

missing translation for 'validMainframeMsg'

Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes.

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Features:

  • Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules that remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface:

  • Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.

TRUSTED_SUSTAINABILITY

Specifications

Binding Property Slightly Hydrophilic
Certified No
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 100 Boxes
Binding Type Adsorption
Color Clear
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 280 μL
Surface Treatment Immulon 2 HB
Well Shape Flat Bottom (F-well)
No. per Case 100
No. per Pack 100
Target Molecule Proteins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.