Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Clear Standard Modules

Catalog No. 12565122 Shop All Thermo Scientific Products
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
PolySorp
Volume (Metric) Working:
250 μL
330 μL
350 μL
Well Shape:
Round
Round, C Bottom
Round, Flat Bottom (F-well)
Well Design:
C12
C8
F12
F16 (2x8 wells)
F8
U8
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Volume (Metric) Working Well Shape Well Design
12-565-122 PolySorp 350 μL Round, Flat Bottom (F-well) F8
22-047029 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-567-130 MaxiSorp 250 μL Round, C Bottom C8
12-565-265 MaxiSorp 250 μL Round, C Bottom C8
12-565-119 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-121 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-127 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-129 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-130 PolySorp 350 μL Round, Flat Bottom (F-well) F8
12-565-123 MaxiSorp 250 μL Round, C Bottom C12
12-565-117 MaxiSorp 250 μL Round U8
12-565-118 PolySorp 250 μL Round U8
14-245-81 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
14-245-167 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
14-245-193B Immulon 4 HBX 330 μL Round, Flat Bottom (F-well) F12
21-377-254 Enhanced Binding 350 μL Round, Flat Bottom (F-well) F8
14-245-80 Immulon 1 B 330 μL Round, Flat Bottom (F-well) F12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 12-565-122 Supplier Thermo Scientific™ Supplier No. 469078
Get 60% Off Storage & Assay Plates
Use promo code PLATE50
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Certifications

Products specified in chart are certified for: Adsorption of IgG; CV of <5% between wells; all results +/-10% from mean; and background of all wells +/-0.005 absorbance units from mean.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Binding Property Hydrophobic
Certified No
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Microplate Strip
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 350 μL
Surface Treatment PolySorp
Well Shape Round, Flat Bottom (F-well)
No. per Case 60
No. per Pack 10
Plate Blocking None
Quantity 60/Cs.
Target Molecule Lipids
Well Design F8
Show More Show Less
What frames do you recommend for your Immuno Clear Standard Modules?

We recommend Frames and Accessories for Immuno Breakable Modules, BreakApart frame (Cat. No. 465404). This frame is designed to hold the Immuno Clear Standard Modules.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Which surfaces do you recommend for RIA (radioimmunoassay)?

The choice of plates or modules will depend on the type of RIA you are planning to perform and specific assay conditions. Nunc provides two different surfaces for immunoassays, PolySorp and MaxiSorp.
- MaxiSorp surface is highly charged and should be used if the assay requires quantitative measurement of proteins (antibodies) or molecules with polar groups.
- PolySorp surface should be used if the assay consists of less polar molecules with hydrophobic characteristics.

Which 96-well Immuno plates or modules do you recommend for various applications?

The following list offers a brief description of the features of Nunc products and their specific applications:
- Nunc-Immuno Plates, MaxiSorp surface - These plates are designed for solid phase immuno assays and have a polystyrene surface with high affinity for polar groups and hydrophilic molecules. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Plates, PolySorp surface - These plates have a polystyrene surface which adsorps less polar molecules compared to the MaxiSorp surface and has a high affinity for hydrophobic groups. These 96-well plates are available with flat (F), round (U), or (C) bottom-well designs.
- Nunc-Immuno Modules - These modules are designed for solid-phase immuno assays. The modules are available in 8-, 12- or 16-well formats with (F), (U), and (C) bottom-well and 8-well BreakApart with (C) bottom-well designs. These different formats allow one to choose the style which is appropriate for their assay design.
BreakApart Modules and LockWell Modules can be used for radioimmunoassays.
- Nunc StarWell Immuno Breakable Modules - The 8-well modules feature eight fins on the inner wall of the (C) bottom wells. This design increases the surface area by 50%. The increase in surface area allows more molecules to be immobilized, increasing assay signal. The fin configuration provides shorter diffusion distance to the surface, reducing incubation times.
- LockWell Immuno Breakable Modules - Each plate consists of 1 x 8 breakable strips. These strips are assembled in a designed frame which locks each well into place by a spring lock. This spring lock design orientates each well at the same horizontal level allowing uniform washing and reading. The LockWell Modules are available with round (U), (C) or StarWell bottom-well designs.
- Plates and Modules with (CovaLink) Covalent Binding Surfaces: These plates can be used to bind proteins, peptides, DNA or carbohydrates. Covalent linkage occurs via secondary amine group. This allows a specific orientation and provides improved stability compared to passive adsorption due to chemical binding between molecules. Binding of molecules on PolySorp and MaxiSorp surfaces is obtained by passive adsorption. CovaLink is a surface grafted with secondary amino groups (<NH) which serves as a bridge for covalent coupling. The covalent binding can immobilize small molecules such as biotin or peptides which may otherwise bind weakly by physical adsorption and facilitate the recognition by detection molecule.
- Nunc FluoroNunc/LumiNunc 96-Well Plates: These plates are optimized for IFMA (Immunofluorometric Assays) or FIA (Fluorometric Immuno Assays). The transparent polystyrene plates give a low background fluorescence and are optimal where a read-through system is used. The white plates/modules provide maximum reflection of fluorescence signal while maintaining low background. The white plates/modules are often used for epifluorescence reading. Black modules reduce background fluorescence and minimize back-scatter light which is often encountered in epifluorescence.

What is the maximum binding capacity for proteins on the Thermo Scientific MaxiSorp surface and PolySorp surface?

Molecules bind to our PolySorp and MaxiSorp surfaces through passive adsorption.
Using IgG as a reference molecule and knowing that it is a globular molecule, theoretical calculations indicate that the maximum binding for proteins are:
- For the MaxiSorp surface, in monolayer: 650 ng/cm^2
- For the PolySorp surface: 220 ng/cm^2.

What is the difference between your Thermo Scientific PolySorp, MaxiSorp, and MiniSorp surfaces?

MaxiSorp, PolySorp and MiniSorp surfaces were developed for immunology assays:
- The MaxiSorp surface is a modified, highly charged polystyrene surface with high affinity for molecules with polar or hydrophilic groups. The surface has a high binding capacity for proteins, including globular antibodies in proper orientation. Thus, it offers very high sensitivity in double antibody "sandwich" tests.
- The PolySorp surface is more hydrophobic than the MaxiSorp surface. It has high affinity to molecules of a more hydrophobic character. This surface is particularly suited to non-protein antigens including virus antigens.
- The MiniSorp surface is a polyethylene surface with very low affinity to molecules of any type. This type of surface is ideal for the liquid phase immuno techniques.
- The MiniSorp surface is only available in a tube format, while the MaxiSorp and PolySorp surfaces are offered in both 96-well plate and module formats.

Which Nunc immunodiagnostic passive surface should I choose: PolySorp, MediSorp, MaxiSorp, or MultiSorp?

Our passive immunodiagnostic surfaces are based on a series of treatments that modify the polystyrene surface to different final levels of hydrophilicity. Selection of one of these surfaces for ELISA development should be based on the characteristics of the biomolecule you seek to immobilize. PolySorp is fairly hydrophobic and should be used for biomolecules that have hydrophobic domains, e.g., lipids, lipoproteins, and large proteins. MediSorp is slightly hydrophilic and can be used for biomolecules with hydrophilic and hydrophobic properties, e.g., medium-to-large proteins such as albumin, or amphiphilic biomolecules such as LPS. MaxiSorp is quite hydrophilic, optimized for binding of IgG (and any other molecule with similar characteristics) and thus is a popular choice for sandwich ELISA and similar protocols. MultiSorp is very hydrophilic, and in turn will immobilize very hydrophilic biomolecules such as glycoproteins. Some optimization will always be necessary, and in many cases screening for binding optimization may involve a few surface types.

What is the difference between a certified and an uncertified immunodiagnostic plate, e.g., Cat. No. 439454 vs. 442404?

There is no difference in the manufacturing process. Certified plates are performance tested (using an antibody binding protocol) during quality control, whereas uncertified plates are not. This test is expensive for us to perform, so we offer the uncertified version at cost savings customers for whom antibody binding testing results are not critical.


Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.