Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Clear Standard Modules
GSA_VA

Catalog No. 12565265
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
PolySorp
Volume (Metric) Working:
250 μL
330 μL
350 μL
Well Shape:
Round
Round, C Bottom
Round, Flat Bottom (F-well)
Well Design:
C12
C8
F12
F16 (2x8 wells)
F8
U8
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Volume (Metric) Working Well Shape Well Design
12-565-265 MaxiSorp 250 μL Round, C Bottom C8
22-047029 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-567-130 MaxiSorp 250 μL Round, C Bottom C8
12-565-119 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-121 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-122 PolySorp 350 μL Round, Flat Bottom (F-well) F8
12-565-127 MaxiSorp 350 μL Round, Flat Bottom (F-well) F16 (2x8 wells)
12-565-129 MaxiSorp 350 μL Round, Flat Bottom (F-well) F8
12-565-130 PolySorp 350 μL Round, Flat Bottom (F-well) F8
12-565-123 MaxiSorp 250 μL Round, C Bottom C12
12-565-117 MaxiSorp 250 μL Round U8
12-565-118 PolySorp 250 μL Round U8
14-245-81 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
14-245-167 Immulon 2 HB 330 μL Round, Flat Bottom (F-well) F12
14-245-193B Immulon 4 HBX 330 μL Round, Flat Bottom (F-well) F12
21-377-254 Enhanced Binding 350 μL Round, Flat Bottom (F-well) F8
14-245-80 Immulon 1 B 330 μL Round, Flat Bottom (F-well) F12
12-565-124 PolySorp 250 μL Round, C Bottom C12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 12-565-265 Supplier Thermo Scientific™ Supplier No. 445101
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat-bottom, C-bottom (radius edge), U-shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.

Certifications

Products specified in chart are certified for: Adsorption of IgG; CV of <5% between wells; all results +/-10% from mean; and background of all wells +/-0.005 absorbance units from mean.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart No
Binding Property Hydrophilic
Certifications/Compliance Certified for adsorption of IgG; CV of <5% between wells
Certified Yes
Format Strip Plate
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 60 Cases
Color Clear
Product Type Strip Plate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Volume (Metric) Working 250 μL
Surface Treatment MaxiSorp
Well Shape Round, C Bottom
No. per Case 60
No. per Pack 10
Plate Blocking None
Quantity 60/Cs.
Target Molecule Antibodies (Glycoproteins)
Well Design C8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.