Learn More
Electron Microscopy Sciences EMS Immuno Mount DAPI & DABCO (IFMDD) Mounting Media 20 ML
SDP
Catalog No. 501927438
Encompass_Preferred
EMS Immuno Mount with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous non-permanent low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes. This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI. Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC). Reagent: Ready to use mounting medium for Immunofluorescence
Catalog No. 50-192-7438 Supplier Electron Microscopy Sciences Supplier No. 1798997
EMS Immuno Mount with anti-fading agent 1, 4-Diazobicyclo-(2,2,2-octane (DABCO) is an aqueous non-permanent low viscosity mounting medium for immunofluorescence. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine and Redox. The fluorescence is retained during storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes. This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI. Not recommended for phycoethyrin (PE), phycocyanin (PC) and allophycocyanin (APC). Reagent: Ready to use mounting medium for Immunofluorescence