Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Immuno Mount w/ Para Phenylenediamine anti fading agent 100 ML
SDP

Catalog No. 501927437
Encompass_Preferred

EMS Immuno Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark. This medium contains phenylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC).

Application: Immunofluorescence, confocal microscopy
Reagent: Ready to use mounting medium with dark coffee color. This color does not interfere with Immunofluorescence.
Refractive Index: 1.400 +/- 0.002 (These numbers apply to these mounting mediums in solution).

Catalog No. 50-192-7437 Supplier Electron Microscopy Sciences Supplier No. 1798981
May include imposed supplier surcharges.
Add to Cart
Add to Cart

EMS Immuno Mount with strong anti-fading agent, 1, 4-phenylenediamine (PPD) is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Alexa fluoro 488, Alexa fluoro 594, tetramethyly rhodamine, and Redox. The fluorescence is retained during prolonged storage at 4 degree C in the dark. This medium contains phenylenediamine and is not suitable for immunofluorescence of Cy dyes, Phycoerythrin (R-PE), phyocyanin (PC), and allophycocyanin (APC).

Application: Immunofluorescence, confocal microscopy
Reagent: Ready to use mounting medium with dark coffee color. This color does not interfere with Immunofluorescence.
Refractive Index: 1.400 +/- 0.002 (These numbers apply to these mounting mediums in solution).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.