Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ ImmunO™ Prostate Specific Antigen (PSA)

Catalog No. MP08322601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08322601 100μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08322601 Supplier MP Biomedicals™ Supplier No. 08322601
Add to Cart
Add to Cart

Filter sterilized

Sourced from human seminal fluid.

  • Purity: >96% by SDS-PAGE
  • Activity: 0.5-5mg PSA/ml
  • pH:7.5
  • Concentration:1mg/ml
  • pI: 6.2-6.5
  • Supplied as solution in 0.05M sodium phosphate buffer containing 0.15M NaCl and 0.09% sodium azide
  • Found negative for HIV 1 and 2 antibodies, Hepatitis B surface antigen and HCV antibodies
  • Sodium azide is added as preservative
  • This product is a biohazard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) Cell Biology and Analysis
Name ImmunO™ Prostate Specific Antigen
pH Range 7.5
Quantity 100μg
Source Human Seminal Fluid
Storage Requirements 0° to 5°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.