Learn More
Thermo Scientific™ Immuno Standard Modules White and Black

Description
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
- Available in 8-, 12- or 16-well strips
- Framed or unframed for choice of optimal assay
- Flat-bottom or C-shaped wells
- Choice of three surface types for optimal binding
- White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
- Black format to reduce backscatter light, which may be encountered in epifluorescence
- Transparent or clear format for use in time-resolved fluorescence applications only
Choice of Surface:
- Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
- Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Breakapart | No |
| Binding Property | Hydrophilic |
| Certified | Yes |
| Format | Standard |
| Detection Method | Fluorescence |
| Array | 8 x 12 |
| Packaging | 60 Cases |
| Binding Type | Adsorption |
| Color | Black |
| Product Type | Microplate Strip |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.