Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Immuno Standard Modules White and Black

Catalog No. 14245196M Shop All Thermo Scientific Products
Change view
Click to view available options
Color:
Black
White
Volume (Metric) Well:
350 μL
380 μL
400 μL
Surface Treatment:
Hydrophobic
MaxiSorp
PolySorp
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric) Well Surface Treatment
14-245-196M White 380 μL Hydrophobic
12-565-289A White 350 μL MaxiSorp
12-565-289B White 350 μL PolySorp
12-565-274 Black 400 μL MaxiSorp
12-565-275 Black 400 μL PolySorp
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-245-196M Supplier Thermo Scientific™ Supplier No. 7566
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.

Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

TRUSTED_SUSTAINABILITY

Specifications

Breakapart Breakable Strips
Binding Property Hydrophobic
Certified No
Format Strip Plate
Lid Without Lid
Detection Method Chemiluminescence
Array 1 x 12
Packaging 320 Pk., 320 Cases
Binding Type Adsorption
Color White
Product Type Microplate Strip
Material Polystyrene
Volume (Metric) Well 380 μL
No. of Wells 96
Volume (Metric) Working 330 μL
Surface Treatment Hydrophobic
Well Shape Round, Flat Bottom (F-well)
For Use With (Application) Luminescence
No. per Case 320
No. per Pack 320
Plate Blocking None
Target Molecule Lipids
Show More Show Less
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.