Learn More
Beckman Coulter IMMUNO-TROL Cells, 60 Tests, CE, IVD
Catalog No. NC1908936
Encompass_Preferred
IMMUNO-TROL Cells are a positive process control for flow cytometry. They are assayed for lymphocyte, granulocyte and monocyte specific antigens and single platform absolute counts. Light scatter, population distribution, fluorescence intensity, and antigen density mimic those of whole blood.
Catalog No. NC1908936 Supplier Beckman Coulter Supplier No. 6607077
IMMUNO-TROL Cells are a positive process control for flow cytometry. They are assayed for lymphocyte, granulocyte and monocyte specific antigens and single platform absolute counts. Light scatter, population distribution, fluorescence intensity, and antigen density mimic those of whole blood.