Learn More
ANP Technologies Inc Immunoassay Diluent, Ready-to-use, PK, IM, direct antigen detection, High signal range, Low background signal, Volume 1 L, pH 7.4, ELISA compatible with HRP, Lateral flow assay compatible
SDP
Catalog No. NC2306274
Encompass
Immunoassay Diluent, Ready-to-use, PK, IM, direct antigen detection, High signal range, Low background signal, Volume 1 L, pH 7.4, ELISA compatible with HRP, Lateral flow assay compatible
Catalog No. NC2306274 Supplier ANP Technologies Inc Supplier No. 9010011L
Immunoassay Diluent, Ready-to-use, PK, IM, direct antigen detection, High signal range, Low background signal, Volume 1 L, pH 7.4, ELISA compatible with HRP, Lateral flow assay compatible