Learn More
- Home
- Products
- Research and Clinical Diagnostic Tests and Controls
- Allergy, Arthritis and Immunology Testing
- Whole Allergen Extracts
- Thermo Scientific™ ImmunoCAP™ Epidermal and Animal Protein Allergens
Thermo Scientific™ ImmunoCAP™ Epidermal and Animal Protein Allergens
Catalog No. 14439601P| Catalog No. | Quantity | Code | Common Name | Regulatory Status |
|---|---|---|---|---|
| 14-439-601P | 10 Tests | e73 | Rat epithelium | FDA Cleared |
| 14-411-001P | 16 Tests | e5 | Dog dander | FDA Cleared |
| 14-410-901P | 16 Tests | e1 | Cat dander | FDA Cleared |
| 14-412-201P | 16 Tests | e3 | Horse dander | FDA Cleared |
| 14-416-601P | 16 Tests | e4 | Cow dander | FDA Cleared |
| 14-416-701P | 16 Tests | e6 | Guinea pig epithelium | FDA Cleared |
| 14-416-901P | 16 Tests | e70 | Goose feathers | FDA Cleared |
| 14-417-001P | 10 Tests | e85 | Chicken feathers | FDA Cleared |
| 14-417-101P | 10 Tests | e86 | Duck feathers | FDA Cleared |
| 14-430-401P | 10 Tests | e78 | Budgerigar feathers | FDA Cleared |
| 14-430-801P | 10 Tests | e82 | Rabbit epithelium | FDA Cleared |
| 14-430-301P | 10 Tests | e77 | Budgerigar droppings | FDA Cleared |
| 14-430-701P | 10 Tests | e81 | Sheep epithelium | FDA Cleared |
| 14-430-901P | 10 Tests | e83 | Swine epithelium | FDA Cleared |
| 14-430-601P | 10 Tests | e80 | Goat epithelium | FDA Cleared |
| 14-431-001P | 10 Tests | e84 | Hamster epithelium | FDA Cleared |
| 14-432-601P | 10 Tests | e87 | Rat epithelium, serum proteins and urine proteins | FDA Cleared |
| 14-432-701P | 10 Tests | e88 | Mouse epithelium, serum proteins and urine proteins | FDA Cleared |
| 14-435-601P | 10 Tests | e89 | Turkey feathers | FDA Cleared |
| 14-439-401P | 10 Tests | e71 | Mouse epithelium | FDA Cleared |
| 14-439-501P | 10 Tests | e72 | Mouse urine proteins | FDA Cleared |
| 14-439-701P | 10 Tests | e74 | Rat urine proteins | FDA Cleared |
| 14-439-901P | 10 Tests | e76 | Mouse serum proteins | FDA Cleared |
| 14-439-801P | 10 Tests | e75 | Rat serum proteins | FDA Cleared |
| 14-482-401P | 10 Tests | e201 | Canary bird feathers | FDA Cleared |
| 14-501-310P | 10 Tests | Re214 | Finch feathers | ASR |
| 14-501-510P | 10 Tests | Re209 | Gerbil epithelium | ASR |
| 14-503-410P | 10 Tests | Re211 | Rabbit, urine proteins | ASR |
| 14-503-210P | 10 Tests | Re215 | Pigeon feathers | ASR |
| 14-503-310P | 10 Tests | Re206 | Rabbit, serum proteins | ASR |
| 14-515-710P | 10 Tests | Re217 | Ferret epithelium | ASR |
| 14-523-010P | 10 Tests | Re219 | Chicken, serum proteins | ASR |
| 14-522-910P | 10 Tests | Re218 | Chicken droppings | ASR |
| 14-416-801P | 16 Tests | e7 | Pigeon droppings | FDA Cleared |
Description
Quantify specific IgE antibodies with ImmunoCAP™ Epidermal and Animal Protein Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
- Truly quantitative measuring range from 0.1 to 100 kUA/L
- Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
- Excellent consistency over time, and between countries, systems, labs and persons
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
- Specimen collection and preparation
- Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
- Collect blood samples using standard procedures
- Keep specimens at room temperature (RT) for shipping purposes only
- Store at 2–8°C up to one week, otherwise store at –20°C
- Avoid repeated freezing and thawing
Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.
The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.
Knowing IgE antibody levels provides guidance to clinicians to:
- Identify offending allergens
- Tailor advice to individual patients
- Indicate development of tolerance (food allergy, specific immunotherapy)
- Inform optimized individual medical treatment plans
- Refer to specialists appropriately
- Evaluate whether or not specific immunotherapy is an option
Specific IgE testing also aids in the identification of patients at risk of:
- The allergy march – a progression of skin symptoms to respiratory symptoms
- Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
- Progression of recurrent symptoms to persistent symptoms
As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Specifications
| Regulatory Status | FDA Cleared |
| Code Type | Animal Protein |
| Common Name | Rat epithelium |
| Quantity | 10 Tests |
| Code | e73 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.