Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Mold and Yeast Allergens

Catalog No. 14410601P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rm201
Rm204
Rm208
Rm209
Rm211
Rm226
Rm46
m1
m10
m11
m12
m13
m14
m15
m16
m2
m202
m205
m207
m3
m4
m5
m6
m7
m70
m8
m80
m81
m9
Common Name:
Acremonium kiliense
Alternaria alternata
Aspergillus fumigatus
Aspergillus niger
Aureobasidium pullulans
Bipolaris spicifera
Botrytis cinerea
Candida albicans
Chaetomium globosum
Cladosporium herbarum
Curvularia lunata
Epicoccum purpurascens
Fusarium proliferatum (F. moniliforme)
Mucor racemosus
Penicillium chrysogenum (P. notatum)
Penicillium glabrum
Phoma betae
Pityrosporum orbiculare (syn. Malassezia)
Rhizopus nigricans
Setomelanomma rostrata (Helminthosporium halodes)
Staphylococcal enterotoxin A
Staphylococcal enterotoxin B
Staphylococcal enterotoxin TSST
Stemphylium herbarum (S. botryosum)
Tilletia tritici
Trichoderma viride
Trichophyton mentagrophytes var. interdigitale
Trichophyton rubrum
Ulocladium chartarum
Regulatory Status:
ASR
FDA Cleared
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name Regulatory Status
14-410-601P 16 Tests m6 Alternaria alternata FDA Cleared
14-410-501P 16 Tests m2 Cladosporium herbarum FDA Cleared
14-411-901P 16 Tests m3 Aspergillus fumigatus FDA Cleared
14-412-001P 16 Tests m5 Candida albicans FDA Cleared
14-414-501P 16 Tests m4 Mucor racemosus FDA Cleared
14-416-001P 16 Tests m1 Penicillium chrysogenum (P. notatum) FDA Cleared
14-425-801P 16 Tests m7 Botrytis cinerea FDA Cleared
14-426-001P 16 Tests m9 Fusarium proliferatum (F. moniliforme) FDA Cleared
14-425-901P 16 Tests m8 Setomelanomma rostrata (Helminthosporium halodes) FDA Cleared
14-426-101P 16 Tests m10 Stemphylium herbarum (S. botryosum) FDA Cleared
14-426-201P 16 Tests m11 Rhizopus nigricans FDA Cleared
14-426-401P 16 Tests m13 Phoma betae FDA Cleared
14-426-301P 16 Tests m12 Aureobasidium pullulans FDA Cleared
14-426-601P 16 Tests m15 Trichoderma viride FDA Cleared
14-426-501P 16 Tests m14 Epicoccum purpurascens FDA Cleared
14-426-701P 16 Tests m16 Curvularia lunata FDA Cleared
14-448-301P 16 Tests m205 Trichophyton rubrum FDA Cleared
14-483-001P 10 Tests m202 Acremonium kiliense FDA Cleared
14-483-101P 10 Tests m207 Aspergillus niger FDA Cleared
14-488-901P 10 Tests m80 Staphylococcal enterotoxin A FDA Cleared
14-489-001P 10 Tests m81 Staphylococcal enterotoxin B FDA Cleared
14-494-010P 10 Tests Rm46 Bipolaris spicifera ASR
14-502-910P 10 Tests Rm201 Tilletia tritici ASR
14-502-810P 10 Tests Rm204 Ulocladium chartarum ASR
14-520-210P 10 Tests Rm208 Chaetomium globosum ASR
14-520-310P 10 Tests Rm209 Penicillium glabrum ASR
14-522-410P 10 Tests Rm211 Trichophyton mentagrophytes var. interdigitale ASR
14-530-110P 10 Tests Rm226 Staphylococcal enterotoxin TSST ASR
14-434-901P 10 Tests m70 Pityrosporum orbiculare (syn. Malassezia) FDA Cleared
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
This item is not returnable. View return policy
Catalog No. 14-410-601P Supplier Thermo Scientific™ Supplier No. 14410601
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Mold and Yeast Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Regulatory Status FDA Cleared
Code Type Mold/Yeast
Common Name Alternaria alternata
Quantity 16 Tests
Code m6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.