Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ImmunoCAP™ Seed, Legume and Nut Allergens

Catalog No. 14421201P
Change view
Click to view available options
Quantity:
10 Tests
16 Tests
Code:
Rf182
Rf224
Rf226
Rf227
Rf253
Rf287
Rf305
Rf315
Rf316
Rf333
Rf335
Rf345
Rf347
f10
f11
f12
f13
f14
f15
f17
f18
f20
f201
f202
f203
f235
f256
f299
f309
f36
f4
f5
f55
f56
f6
f7
f79
f8
f9
Common Name:
Almond
Barley
Brazil nut
Buckwheat
Cashew nut
Chick pea
Coconut
Common millet
Fenugreek
Foxtail millet
Gluten
Green bean
Hazel nut
Lentil
Lima bean
Linseed
Lupin seed
Macadamia nut
Maize, Corn
Oat
Pea
Peanut
Pecan nut
Pine nut, pignoles
Pistachio
Poppy seed
Pumpkin seed
Quinoa
Rape seed
Red kidney bean
Rice
Rye
Sesame seed
Soybean
Sugar-beet seed
Sweet chestnut
Walnut
Wheat
White bean
39 product options available for selection
Product selection table with 39 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Code Common Name
14-421-201P 16 Tests f36 Coconut
14-411-501P 16 Tests f14 Soybean
14-411-301P 16 Tests f4 Wheat
14-412-601P 16 Tests f13 Peanut
14-412-501P 16 Tests f11 Buckwheat
14-412-401P 16 Tests f8 Maize, Corn
14-412-301P 16 Tests f5 Rye
14-417-201P 16 Tests f6 Barley
14-417-301P 16 Tests f7 Oat
14-417-601P 16 Tests f12 Pea
14-417-401P 16 Tests f9 Rice
14-417-501P 16 Tests f10 Sesame seed
14-417-701P 16 Tests f15 White bean
14-417-801P 16 Tests f18 Brazil nut
14-417-901P 16 Tests f20 Almond
14-429-101P 10 Tests f79 Gluten
14-445-901P 16 Tests f55 Common millet
14-446-001P 16 Tests f56 Foxtail millet
14-452-101P 16 Tests f256 Walnut
14-452-801P 16 Tests f17 Hazel nut
14-480-701P 10 Tests f201 Pecan nut
14-481-501P 10 Tests f235 Lentil
14-483-601P 10 Tests f203 Pistachio
14-483-501P 10 Tests f202 Cashew nut
14-484-301P 10 Tests f299 Sweet chestnut
14-492-910P 10 Tests Rf182 Lima bean
14-509-310P 10 Tests Rf253 Pine nut, pignoles
14-509-710P 10 Tests Rf224 Poppy seed
14-510-010P 10 Tests Rf287 Red kidney bean
14-510-210P 10 Tests Rf227 Sugar-beet seed
14-509-810P 10 Tests Rf226 Pumpkin seed
14-518-710P 10 Tests Rf305 Fenugreek
14-519-601P 10 Tests f309 Chick pea
14-521-110P 10 Tests Rf315 Green bean
14-521-310P 10 Tests Rf316 Rape seed
14-525-010P 10 Tests Rf333 Linseed
14-525-410P 10 Tests Rf335 Lupin seed
14-528-210P 10 Tests Rf345 Macadamia nut
14-528-410P 10 Tests Rf347 Quinoa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
39 options
This item is not returnable. View return policy
Catalog No. 14-421-201P Supplier Thermo Scientific™ Supplier No. 14421201
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.

Quantify specific IgE antibodies with ImmunoCAP™ Seed, Legume and Nut Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.

ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
TRUSTED_SUSTAINABILITY

Specifications

Scientific Name Cocos nucifera
Regulatory Status FDA Cleared
Code Type Seed, Legume, and Nut
Common Name Coconut
Quantity 16 Tests
Code f36
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.